Stampa…
Solid supported liquid/liquid extraction cartridges, HyperSep™
Fornitore: Thermo Fisher Scientific
60109-1000-6-7 60109-20000607 60109-2000-127 60109-200-3-7 60109-4000-257 60109-500-3-7 60109-500-6-7
549-0752EA 171 EUR
549-0752 549-0753 549-0754 549-0755 549-0756 549-0757 549-0758
Solid supported liquid/liquid extraction cartridges, HyperSep™
Colonne cromatografiche
Solid supported liquid/liquid extraction (SLE) is a fast effective sample preparation technique that provides considerable benefits over liquid/liquid extraction (LLE) protocols for removal of phospholipids from biological samples.
  • Greater reproducibility and recoveries compared to LLE techniques
  • Prevents emulsification often associated with LLE as the sample and water immiscible solvents are not in direct contact
  • Reduced solvent requirements compared to LLE
  • Can be completely automated unlike LLE
  • Improved cleanliness of sample extract compared to protein precipitation techniques
  • Improved sensitivity compared to protein precipitation techniques