Stampa…
Crimp neck vials, N 8
Fornitore: MACHEREY-NAGEL
70282 70286
MANA70282EA 19.5 EUR
MANA70282 MANA70286
Crimp neck vials, N 8
Vial
These crimp neck vials are suitable for micro sampling.
  • Made of clear and amber glass
  • Available with flat, round or conical bottom
Useable with adapter for the common autosamplers of Agilent, CTC, Dionex (Thermo Scientific), Knauer, PerkinElmer, Shimadzu, Thermo Scientific, Varian (Agilent), VWR® (Merck / Hitachi), etc.
Ready-to-use crimp caps with assembled septa are available for the crimp neck vials.
- High purity silicone/PTFE septa for more demanding analyses
- Economic natural rubber/butyl/TEF or Red Rubber/FEP septa