Potassium tetraoxalate dihydrate ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Oxalic acid, potassium salt dihydrate, Potassium trihydrogen dioxalate dihydrate
P0475-25G
P0475-500G
TCIAP0475-25GEA
16
EUR
TCIAP0475-25G
TCIAP0475-500G
Potassium tetraoxalate dihydrate ≥99.0% (by titrimetric analysis)
Potassium tetraoxalate dihydrate
Formula:
C₄H₃KO₈·2H₂O Peso molecolare: 254,19 g/mol (218,16 anhy) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00036364 CAS Number: 6100-20-5 EINECS: 204-874-6 UN: 2811 ADR: 6.1,III Merck Index: 14,07689 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Redox method) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...