2-Biphenylyl isocyanate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Biphenyl Isocyanate, Isocyanic acid 2-Biphenyl ester, 2-Biphenylylisocyanate
I0503-25G
I0503-5G
TCIAI0503-25GEA
361
EUR
TCIAI0503-25G
TCIAI0503-5G
2-Biphenylyl isocyanate ≥98.0% (by GC)
2-Biphenylyl isocyanate
Formula:
C₁₃H₉NO Peso molecolare: 195,2167 g/mol Punto di ebollizione: 130°C |
MDL Number:
MFCD00037087 CAS Number: 17337-13-2 UN: 2206 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...