Stampa…

2',7'-Dichlorofluorescein ≥95.0% (by HPLC) fluorescent indicator

Fornitore: TCI
Attenzione

D0371-1G D0371-25G
TCIAD0371-1GEA 16 EUR
TCIAD0371-1G TCIAD0371-25G
2',7'-Dichlorofluorescein ≥95.0% (by HPLC) fluorescent indicator
2',7'-Dichlorofluorescein
Formula: C₂₀H₁₀Cl₂O₅
MDL Number: MFCD00005047
CAS Number: 76-54-0
EINECS: 200-968-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27557e5db648ba217ad45ef92a16f0291f36e76d&vid=c746e7ea4382b3583c1333d74b0b359b0448170a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Drying loss max. 10.0 %
Sensitiveness to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR