Stampa…

(6-Chloro-3-pyridinyl)methanol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine

C1897-25G C1897-5G
TCIAC1897-25GEA 210 EUR
TCIAC1897-25G TCIAC1897-5G
(6-Chloro-3-pyridinyl)methanol ≥98.0% (by GC)
(6-Chloro-3-pyridinyl)methanol
Formula: C₆H₆ClNO
Peso molecolare: 143,57 g/mol
MDL Number: MFCD01075226
CAS Number: 21543-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=397c0fc79fd296bd5061535df9331bf8969a3191&vid=397c0fc79fd296bd5061535df9331bf8969a3191&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR