Stampa…

Propargyltriphenylphosphonium bromide ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

P1438-25G P1438-5G
TCIAP1438-25GEA 126 EUR
TCIAP1438-25G TCIAP1438-5G
Propargyltriphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Propargyltriphenylphosphonium bromide
Formula: C₂₁H₁₈BrP
MDL Number: MFCD00011811
CAS Number: 2091-46-5
EINECS: 218-242-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31a5448a3e52969d74b8e10831a143182c5c81f3&vid=31a5448a3e52969d74b8e10831a143182c5c81f3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR