Stampa…

(R)-(-)-2-Oxothiazolidine-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: (R)-2-Oxothiazolidine-4-carboxylic acid, Procysteine

T1223-1G T1223-5G
TCIAT1223-1GEA 27 EUR
TCIAT1223-1G TCIAT1223-5G
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid ≥97.0% (by HPLC, titration analysis)
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
Formula: C₄H₅NO₃S
MDL Number: MFCD00066092
CAS Number: 19771-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=601f660f9a05c22701d532f82cdfca331a7d36a2&vid=601f660f9a05c22701d532f82cdfca331a7d36a2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -61.0 to -66.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR