Stampa…

Thiophene-2-carbonyl chloride ≥99.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: 2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride

T0177-25G T0177-500G T0177-100G
TCIAT0177-25GEA 34 EUR
TCIAT0177-25G TCIAT0177-500G TCIAT0177-100G
Thiophene-2-carbonyl chloride ≥99.0% (by titrimetric analysis)
Thiophene-2-carbonyl chloride
Formula: C₅H₃ClOS
Peso molecolare: 146,6 g/mol
Punto di ebollizione: 190°C
Flash Pt: 90°C
MDL Number: MFCD00005428
CAS Number: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aac595de6b2d136dc1f23129b894f48157db4ae&vid=5aac595de6b2d136dc1f23129b894f48157db4ae&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Brown clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR