Stampa…

1-Ethyl-2,3-piperazinedione ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Ethylpiperazine-2,3-dione

E0459-25G E0459-250G
TCIAE0459-25GEA 109 EUR
TCIAE0459-25G TCIAE0459-250G
1-Ethyl-2,3-piperazinedione ≥98.0% (by GC)
1-Ethyl-2,3-piperazinedione
Formula: C₆H₁₀N₂O₂
Peso molecolare: 142,16 g/mol
MDL Number: MFCD00051825
CAS Number: 59702-31-7
EINECS: 261-866-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a89232d40060c95e0fd8117f64d6ad2003ff5098&vid=a89232d40060c95e0fd8117f64d6ad2003ff5098&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR