Stampa…

1,3-Butanediol dimethacrylate ≥98.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,3-Butylene glycol dimethacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

B1065-25ML B1065-500ML
TCIAB1065-25MLEA 16 EUR
TCIAB1065-25ML TCIAB1065-500ML
1,3-Butanediol dimethacrylate ≥98.0% (by GC) stabilizzato
1,3-Butanediol dimethacrylate
Formula: C₁₂H₁₈O₄
Peso molecolare: 226,27 g/mol
Flash Pt: 130°C
MDL Number: MFCD00014930
CAS Number: 1189-08-8
EINECS: 214-711-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb9b87a3305fae8b47dbff9cfcdff7a1690e043e&vid=eb9b87a3305fae8b47dbff9cfcdff7a1690e043e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR