Stampa…

Sodium dicyanoamide ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Sodium dicyanamide

S0838-25G S0838-100G
TCIAS0838-25GEA 38 EUR
TCIAS0838-25G TCIAS0838-100G
Sodium dicyanoamide ≥98.0% (by titrimetric analysis)
Sodium dicyanoamide
Formula: C₂N₃Na
Peso molecolare: 89,03 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003535
CAS Number: 1934-75-4
EINECS: 217-703-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2991366ac3d840852a18864e44d6fd46e2279272&vid=2991366ac3d840852a18864e44d6fd46e2279272&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR