Phenylboronic acid pinacol ester ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₂H₁₇BO₂ Punto di ebollizione: 130°C |
MDL Number:
MFCD00966985 CAS Number: 24388-23-6 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...