Stampa…

1,2,3-Trichloro-5-nitrobenzene ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Nitro-3,4,5-trichlorobenzene, 5-Nitro-1,2,3-trichlorobenzene

T1580-25G T1580-5G
TCIAT1580-25GEA 106 EUR
TCIAT1580-25G TCIAT1580-5G
1,2,3-Trichloro-5-nitrobenzene ≥96.0% (by GC)
1,2,3-Trichloro-5-nitrobenzene
Formula: C₆H₂Cl₃NO₂
Peso molecolare: 226,45 g/mol
Punto di fusione: 71 °C
Densità: 1,807 g/cm³ (20 °C)
MDL Number: MFCD00061096
CAS Number: 20098-48-0
EINECS: 243-511-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75185241c2c60e8b8920b5e52aab8160a7e60636&vid=4746e81f19d7e6448d00e235b1de9c78b91173ea&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 68.0 to 73.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR