2,4,6-Triallyloxy-1,3,5-triazine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,4,6-Tris(allyloxy)-1,3,5-triazine, 2,4,6-Triallyloxy-s-triazine, 2,4,6-Tris(allyloxy)-s-triazine
T0333-25G
T0333-500G
TCIAT0333-25GEA
16
EUR
TCIAT0333-25G
TCIAT0333-500G
2,4,6-Triallyloxy-1,3,5-triazine ≥98.0% (by GC)
2,4,6-Triallyloxy-1,3,5-triazine
Formula:
C₁₂H₁₅N₃O₃ Peso molecolare: 249,27 g/mol Punto di ebollizione: 162°C |
MDL Number:
MFCD00006049 CAS Number: 101-37-1 EINECS: 202-936-7 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Solubility in EtOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...