Stampa…

Titanyl(IV) acetylacetonate

Fornitore: TCI
Attenzione

Synonyms: Titanium(IV) oxide acetylacetonate, Titanium(IV) oxyacetylacetonate, Titanyl Acetylacetonate, Titanium (IV) oxide bis(acetylacetonate), Oxobis(2,4-pentanedionato-O,O')titanium (IV), Titanium (IV) oxide bis(2,4-pentanedionate)

T0249-25G T0249-500G
TCIAT0249-25GEA 80 EUR
TCIAT0249-25G TCIAT0249-500G
Titanyl(IV) acetylacetonate
Titanyl(IV) acetylacetonate
Formula: C₁₀H₁₄O₅Ti
MDL Number: MFCD00013505
CAS Number: 14024-64-7
EINECS: 237-861-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15766d541ae16499b746cb1c142c01137727bbce&vid=15766d541ae16499b746cb1c142c01137727bbce&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Ignition residue 28.0 to 34.0 % (as TiO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR