Stampa…

3,3'-Diaminodifenilsulfone ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,3'-Sulfonildianilina

B0394-25G B0394-500G
TCIAB0394-25GEA 17 EUR
TCIAB0394-25G TCIAB0394-500G
3,3'-Diaminodifenilsulfone ≥98.0% (by titrimetric analysis)
3,3'-Diaminodifenilsulfone
Formula: C₁₂H₁₂N₂O₂S
Peso molecolare: 248,31 g/mol
MDL Number: MFCD00007792
CAS Number: 599-61-1
EINECS: 209-967-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9069fcd7193128be84f213d4ea08894092d2df0&vid=e9069fcd7193128be84f213d4ea08894092d2df0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 175.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR