Stampa…

Sodium hexadecyl sulphate ≥95.0% (by titrimetric analysis) (contains ca. 40% Sodium Stearyl sulphate)

Fornitore: TCI
Attenzione

H0080-25G H0080-500G
TCIAH0080-25GEA 16 EUR
TCIAH0080-25G TCIAH0080-500G
Sodium hexadecyl sulphate ≥95.0% (by titrimetric analysis) (contains ca. 40% Sodium Stearyl sulphate)
Sodium hexadecyl sulphate
Formula: C₁₆H₃₃NaO₄S
MDL Number: MFCD00047766
CAS Number: 1120-01-0
EINECS: 214-292-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9b06716867bc1e93f4a540c8caefecd7ccd6c44&vid=f2574532eb9555636ceff565859fa0784ca26003&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 50.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR