3-Nitro-1H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis) coupling agent
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Nitro-1,2,4-1H-triazole, 3-Nitro-1,2,4-triazole
N0477-5G
N0477-1G
TCIAN0477-5GEA
98
EUR
TCIAN0477-5G
TCIAN0477-1G
3-Nitro-1H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis) coupling agent
3-Nitro-1H-1,2,4-triazole
Formula:
C₂H₂N₄O₂ Peso molecolare: 114,06 g/mol Punto di fusione: 210…215 °C Densità: 1,65 g/cm³ (20 °C) Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00009749 CAS Number: 24807-55-4 EINECS: 246-468-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...