Stampa…

N,N-Dimetil p-fenilendiammina solfato ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N,N-Dimethyl-1,4-phenylenediammonium sulphate, N,N-Dimethylbenzene-1,4-diammonium sulphate

D0782-500G D0782-25G
TCIAD0782-500GEA 920 EUR
TCIAD0782-500G TCIAD0782-25G
N,N-Dimetil p-fenilendiammina solfato ≥98.0% (by titrimetric analysis)
N,N-Dimetil p-fenilendiammina solfato
Formula: C₈H₁₄N₂O₄S
Peso molecolare: 234,28 g/mol
Punto di fusione: 205°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00012992
CAS Number: 536-47-0
EINECS: 208-636-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59ac8efbbdc97c4f99842d96ad20bfb0fe7c84ba&vid=59ac8efbbdc97c4f99842d96ad20bfb0fe7c84ba&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR