2-Methyl-2-nitropropane-1,3-diol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Methyl-2-nitro-1,3-propanediol, 2-Nitro-2-methyl-1,3-propanediol
M0639-25G
M0639-5G
TCIAM0639-25GEA
156
EUR
TCIAM0639-25G
TCIAM0639-5G
2-Methyl-2-nitropropane-1,3-diol ≥98.0% (by GC)
2-Methyl-2-nitropropane-1,3-diol
Formula:
C₄H₉NO₄ Peso molecolare: 135,12 g/mol Punto di ebollizione: 324 °C (1013 hPa) |
MDL Number:
MFCD00024812 CAS Number: 77-49-6 EINECS: 201-031-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 154.0 to 158.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...