Stampa…

Bis(4-chlorophenyl)disulphide ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Bis(4-chlorophenyl) disulphide

D0360-25G D0360-5G
TCIAD0360-25GEA 102 EUR
TCIAD0360-25G TCIAD0360-5G
Bis(4-chlorophenyl)disulphide ≥98.0% (by GC)
Bis(4-chlorophenyl)disulphide
Formula: C₁₂H₈Cl₂S₂
Peso molecolare: 287,23 g/mol
Punto di ebollizione: 180°C
MDL Number: MFCD00013642
CAS Number: 1142-19-4
EINECS: 214-531-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d3adeb7917798f0dd9e98c24c2adc7c0636869f&vid=6d3adeb7917798f0dd9e98c24c2adc7c0636869f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR