Stampa…

4-Tolylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dihydroxy(4-methylphenyl)borane, p-Tolylboronic acid, 4-Methylphenylboronic acid, (4-Methylphenyl)metaboric acid, 4-Methylbenzeneboronic acid

M1126-25G M1126-5G
TCIAM1126-25GEA 227 EUR
TCIAM1126-25G TCIAM1126-5G
4-Tolylboronic acid (contains varying amounts of Anhydride)
4-Tolylboronic acid
Formula: C₇H₉BO₂
Peso molecolare: 135,96 g/mol
Punto di fusione: 260 °C
MDL Number: MFCD00039138
CAS Number: 5720-05-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88a499cf2271d075c72cab3183ccd4470856d940&vid=88a499cf2271d075c72cab3183ccd4470856d940&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR