2,3,5,6-Tetramethyl-m-phenylenediamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
3,6-Diaminodurene, 2,3,5,6-Tetramethylbenzene-1,3-diamine, 3,6-Diamino-1,2,4,5-tetramethylbenzene, 1,4-Diamino-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-1,4-phenylenediamine, 2,3,5,6-Tetramethyl-1,3-phenylenediamine
Formula:
C₁₀H₁₆N₂ Peso molecolare: 164.25 g/mol Punto di ebollizione: 316 |
MDL Number:
MFCD00007905 CAS Number: 3102-87-2 EINECS: 221-457-4 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...