Stampa…

(S)-(+)-Mandelic acid ≥99.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M0661-25G M0661-250G
TCIAM0661-25GEA 44 EUR
TCIAM0661-25G TCIAM0661-250G
(S)-(+)-Mandelic acid ≥99.0% (by GC, titration analysis)
(S)-(+)-Mandelic acid
Formula: C₈H₈O₃
Punto di ebollizione: 322 °C (1013 hPa)
Punto di fusione: 130…132 °C
Densità: 1,349 g/cm³ (20 °C)
Flash Pt: ≥ 190 °C
MDL Number: MFCD00004495
CAS Number: 17199-29-0
EINECS: 241-240-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=740f275191c4efc81631cb30e369f39124ce93b3&vid=740f275191c4efc81631cb30e369f39124ce93b3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 99.0 %
Melting point 132.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR