Stampa…

(R)-(-)-Methyl mandelate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester

M1349-1G M1349-5G
TCIAM1349-1GEA 20 EUR
TCIAM1349-1G TCIAM1349-5G
(R)-(-)-Methyl mandelate ≥98.0% (by GC)
(R)-(-)-Methyl mandelate
Formula: C₉H₁₀O₃
Peso molecolare: 166,18 g/mol
Punto di ebollizione: 267 °C (1013 hPa)
Punto di fusione: 51…53 °C
MDL Number: MFCD00064247
CAS Number: 20698-91-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=568f5741483ae65461ade5753835d7e0cf09def5&vid=568f5741483ae65461ade5753835d7e0cf09def5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Specific rotation [a]20/D -140 to -150 deg (C=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR