Stampa…

Palladium(II) acetylacetonate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Palladium (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')palladium (II), Palladium (II) bis(2,4-pentanedionato-O,O'), Palladium(II) 2,4-pentanedionate

B2018-1G B2018-5G
TCIAB2018-1GEA 219 EUR
TCIAB2018-1G TCIAB2018-5G
Palladium(II) acetylacetonate ≥98.0% (by titrimetric analysis)
Palladium(II) acetylacetonate
Formula: C₁₀H₁₄O₄Pd
Peso molecolare: 304,64 g/mol
Punto di ebollizione: ? 190 °C (1013 hPa)
Punto di fusione: 190 °C
MDL Number: MFCD00000025
CAS Number: 14024-61-4
EINECS: 237-859-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7ccb094adbdfea59d055bdd02edcee2e332949c&vid=a7ccb094adbdfea59d055bdd02edcee2e332949c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR