Stampa…
α,α,α,5-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 3-Fluoro-5-trifluoromethylbenzoic acid, 3-Fluoro-5-(trifluoromethyl)benzoic acid
F0759-5G F0759-1G
TCIAF0759-5GEA 86 EUR
TCIAF0759-5G TCIAF0759-1G
α,α,α,5-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
α,α,α,5-Tetrafluoro-m-toluic acid
Formula: C₈H₄F₄O₂
Peso molecolare: 208,11 g/mol
MDL Number: MFCD00061293
CAS Number: 161622-05-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18e095dcffad17b6480ac37551026a8de28f4dbd&vid=18e095dcffad17b6480ac37551026a8de28f4dbd&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 102.0 to 106.0 deg-C