Stampa…

2'-Aminoacetanilide ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

A2426-5G A2426-25G
TCIAA2426-5GEA 50 EUR
TCIAA2426-5G TCIAA2426-25G
2'-Aminoacetanilide ≥98.0% (by titrimetric analysis)
2'-Aminoacetanilide
Formula: C₈H₁₀N₂O
MDL Number: MFCD00210388
CAS Number: 34801-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d99c1457277121753279f349fad62c1479cee4d&vid=0d99c1457277121753279f349fad62c1479cee4d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Light red powder to crystalline
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR