(S)-(-)-1-Octyn-3-ol ≥99.0% (by GC) omega side-chain unit for PG synthesis
Fornitore: TCI
|
Attenzione
|
O0235-1G
O0235-5G
TCIAO0235-1GEA
182
EUR
TCIAO0235-1G
TCIAO0235-5G
(S)-(-)-1-Octyn-3-ol ≥99.0% (by GC) omega side-chain unit for PG synthesis
(S)-(-)-1-Octyn-3-ol
Formula:
C₈H₁₄O Peso molecolare: 126,2 g/mol Punto di ebollizione: 130°C Densità: 0,866 g/cm³ (20 °C) Flash Pt: 64°C |
MDL Number:
MFCD00191475 CAS Number: 32556-71-1 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 99.0 % |
Specific rotation [a]20/D | -9.0 to -11.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...