(S)-(-)-1-Phenylethanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol
P0796-1G
P0796-25G
P0796-5G
TCIAP0796-1GEA
47
EUR
TCIAP0796-1G
TCIAP0796-25G
TCIAP0796-5G
(S)-(-)-1-Phenylethanol ≥98.0% (by GC)
(S)-(-)-1-Phenylethanol
Formula:
C₈H₁₀O Peso molecolare: 122,17 g/mol Punto di ebollizione: 98°C Densità: 1,018 g/cm³ (20 °C) Flash Pt: 85°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00064264 CAS Number: 1445-91-6 UN: 2937 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific rotation [a]20/D | -40.0 to -46.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...