D-(-)-Arginine ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
D-Arginine, (R)-2-Amino-5-guanidinopentanoic acid, H-D-Arg-OH, H-D-(-)-Arg-OH, (R)-(-)-Arginine, D-(-)-Arginine, D-2-Amino-5-guanidinovaleric acid
A1029-1G
A1029-5G
TCIAA1029-1GEA
31
EUR
TCIAA1029-1G
TCIAA1029-5G
D-(-)-Arginine ≥98.0% (by titrimetric analysis)
D-(-)-Arginine
Formula:
C₆H₁₄N₄O₂ Peso molecolare: 174,2 g/mol Punto di fusione: 212 °C |
MDL Number:
MFCD00063116 CAS Number: 157-06-2 EINECS: 205-866-5 |
|
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -24.0 to -28.0 deg (C=8, 6mol/L HCl) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...