Stampa…

3-(Indol-3-yl)propionic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

I0032-25G I0032-5G
TCIAI0032-25GEA 73 EUR
TCIAI0032-25G TCIAI0032-5G
3-(Indol-3-yl)propionic acid ≥98.0% (by titrimetric analysis)
3-(Indol-3-yl)propionic acid
Formula: C₁₁H₁₁NO₂
Punto di fusione: 129…132 °C
Densità: 1,356 g/cm³ (20 °C)
MDL Number: MFCD00005660
CAS Number: 830-96-6
EINECS: 212-600-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e6e1ff00284f809045e9bfbf9d95cc93cf3e68e&vid=1e6e1ff00284f809045e9bfbf9d95cc93cf3e68e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR