2-Naphthyl boronic acid (contains varying amounts of Anhydride)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid
N0649-1G
N0649-25G
N0649-5G
TCIAN0649-1GEA
18
EUR
TCIAN0649-1G
TCIAN0649-25G
TCIAN0649-5G
2-Naphthyl boronic acid (contains varying amounts of Anhydride)
2-Naphthyl boronic acid
Formula:
C₁₀H₉BO₂ Peso molecolare: 171,99 g/mol |
MDL Number:
MFCD00236051 CAS Number: 32316-92-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...