Stampa…

Taurina ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Aminoethanesulphonic acid

A0295-25G A0295-500G
TCIAA0295-25GEA 18 EUR
TCIAA0295-25G TCIAA0295-500G
Taurina ≥98.0% (by titrimetric analysis)
Taurina
Formula: C₂H₇NO₃S
Peso molecolare: 125,15 g/mol
Punto di fusione: 300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8
Merck Index: 14,09074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc41fec3cabc37efab37e5367e4a776aa29e67b0&vid=dc41fec3cabc37efab37e5367e4a776aa29e67b0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR