Ethyl indol-3-ylacetate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl 3-indoleacetate, Ethyl 2-(1H-indol-3-yl)acetate, Ethyl 3-Indoleacetate, IAA ethyl ester, Indole-3-acetic acid ethyl ester, Ethyl indole-3-acetate, Indol-3-ylacetic acid ethyl ester, 3-Indoleacetic acid ethyl ester
E0878-25G
E0878-5G
TCIAE0878-25GEA
64
EUR
TCIAE0878-25G
TCIAE0878-5G
Ethyl indol-3-ylacetate ≥98.0% (by GC)
Ethyl indol-3-ylacetate
Formula:
C₁₂H₁₃NO₂ Peso molecolare: 203,24 g/mol Punto di ebollizione: 166°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00005635 CAS Number: 778-82-5 EINECS: 212-296-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...