Stampa…

2-Nitrophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dihydroxy(2-nitrophenyl)borane, (2-Nitrophenyl)boronic acid, 2-Nitrobenzeneboronic acid, (2-Nitrophenyl)metaboric acid

N0811-1G N0811-5G
TCIAN0811-1GEA 58 EUR
TCIAN0811-1G TCIAN0811-5G
2-Nitrophenylboronic acid (contains varying amounts of Anhydride)
2-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
Peso molecolare: 166,93 g/mol
Punto di fusione: 142°C
MDL Number: MFCD00161358
CAS Number: 5570-19-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc328068fc74139da075b8eb8b1d095230223107&vid=cc328068fc74139da075b8eb8b1d095230223107&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR