Stampa…

4-Amino-1-isopropylpiperidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

I0774-25G I0774-5G
TCIAI0774-25GEA 243 EUR
TCIAI0774-25G TCIAI0774-5G
4-Amino-1-isopropylpiperidine ≥98.0% (by GC, titration analysis)
4-Amino-1-isopropylpiperidine
Formula: C₈H₁₈N₂
Peso molecolare: 142.2419 g/mol
MDL Number: MFCD03411606
CAS Number: 127285-08-9
UN: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24073abadecd2de07fb8b6c6f4a31fa996d3e39f&vid=24073abadecd2de07fb8b6c6f4a31fa996d3e39f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR