Stampa…

1-Benzyl-4-hydroxypiperidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 1-Benzylpiperidin-4-ol

B2252-25G B2252-5G
TCIAB2252-25GEA 65 EUR
TCIAB2252-25G TCIAB2252-5G
1-Benzyl-4-hydroxypiperidine ≥98.0% (by GC, titration analysis)
1-Benzyl-4-hydroxypiperidine
Formula: C₁₂H₁₇NO
Peso molecolare: 191.27 g/mol
MDL Number: MFCD00006503
CAS Number: 4727-72-4
EINECS: 225-226-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfcdfa7a2b9705817e033c4f8b65380683bfc109&vid=dfcdfa7a2b9705817e033c4f8b65380683bfc109&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR