Stampa…

1,4-Dichloro-9,10-anthraquinone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,4-Dichloroanthracene-9,10-dione

D2518-1G D2518-5G
TCIAD2518-1GEA 159 EUR
TCIAD2518-1G TCIAD2518-5G
1,4-Dichloro-9,10-anthraquinone ≥98.0% (by GC)
1,4-Dichloro-9,10-anthraquinone
Formula: C₁₄H₆Cl₂O₂
Peso molecolare: 277,1022 g/mol
MDL Number: MFCD00192002
CAS Number: 602-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddd4e0f5fca36caaab5722c8c608dee90e4c7ef5&vid=ddd4e0f5fca36caaab5722c8c608dee90e4c7ef5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR