1-Bromo-2-(bromomethyl)naphthalene ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B1607-1G
B1607-25G
B1607-5G
TCIAB1607-1GEA
56
EUR
TCIAB1607-1G
TCIAB1607-25G
TCIAB1607-5G
1-Bromo-2-(bromomethyl)naphthalene ≥97.0% (by GC)
1-Bromo-2-(bromomethyl)naphthalene
Formula:
C₁₁H₈Br₂ Peso molecolare: 299,99 g/mol |
MDL Number:
MFCD00046369 CAS Number: 37763-43-2 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...