Stampa…

Gadolinium nitride ≥99.5%

Fornitore: Thermo Fisher Scientific

Synonyms: Gadolinium(III) nitride

46709.03 46709.06
46709.03EA 42 EUR
46709.03 46709.06
Gadolinium nitride ≥99.5%
Gadolinium nitride
Formula: GdN
Peso molecolare: 171.26 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00049943
CAS Number: 25764-15-2
EINECS: 247-250-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR