Stampa…
5-Methylisophthalic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Attenzione
M1903-25G M1903-5G
TCIAM1903-25GEA 163 EUR
TCIAM1903-25G TCIAM1903-5G
5-Methylisophthalic acid ≥98.0% (by GC, titration analysis)
5-Methylisophthalic acid
Formula: C₉H₈O₄
Punto di fusione: 298°C
MDL Number: MFCD00013986
CAS Number: 499-49-0
EINECS: 207-881-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a195d40d958aa4c4fe0809309f9e20d2333b6c99&vid=a195d40d958aa4c4fe0809309f9e20d2333b6c99&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %