Stampa…

m-Anisidinium chloride ≥99.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3-Methoxyaniline hydrochloride, m-Anisidine hydrochloride

A0488-25G A0488-5G
TCIAA0488-25GEA 141 EUR
TCIAA0488-25G TCIAA0488-5G
m-Anisidinium chloride ≥99.0% (by HPLC, total nitrogen)
m-Anisidinium chloride
Formula: C₇H₁₀ClNO
Peso molecolare: 159,6100 g/mol
MDL Number: MFCD00060220
CAS Number: 27191-09-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bd6cef5171386f6d2ef3f1e1cbfd9a799f97619&vid=6bd6cef5171386f6d2ef3f1e1cbfd9a799f97619&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR