Stampa…

Isobutyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

H0493-25G H0493-500G H0493-100G
TCIAH0493-25GEA 28 EUR
TCIAH0493-25G TCIAH0493-500G TCIAH0493-100G
Isobutyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis)
Isobutyl-4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
MDL Number: MFCD00020167
CAS Number: 4247-02-3
EINECS: 224-208-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cd9f47d0da18d8ca43c2b02bbb51d7a36530c28&vid=1cd9f47d0da18d8ca43c2b02bbb51d7a36530c28&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 75.0 to 79.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR