Stampa…

sec-Butyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Hydroxybenzoic acid sec-butyl ester

H0496-25G H0496-500G H0496-100G
TCIAH0496-25GEA 73 EUR
TCIAH0496-25G TCIAH0496-500G TCIAH0496-100G
sec-Butyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
sec-Butyl-4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
Punto di ebollizione: 210°C
MDL Number: MFCD00059626
CAS Number: 17696-61-6
EINECS: 241-697-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea33d502a51364e9dc566bc26f3a25ece89d2c8f&vid=ea33d502a51364e9dc566bc26f3a25ece89d2c8f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 59.0 to 62.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR