Stampa…

1-Bromo-4-hexyloxybenzene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-(4-Bromophenoxy)hexane, 1-Bromo-4-(hexyloxy)benzene

B3876-25G B3876-5G
TCIAB3876-25GEA 227 EUR
TCIAB3876-25G TCIAB3876-5G
1-Bromo-4-hexyloxybenzene ≥98.0% (by GC)
1-Bromo-4-hexyloxybenzene
Formula: C₁₂H₁₇BrO
Peso molecolare: 257,18 g/mol
Punto di ebollizione: 154°C
MDL Number: MFCD00173753
CAS Number: 30752-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11853bd7a5a7d9a0fe8fd9448dc4571cd8924975&vid=11853bd7a5a7d9a0fe8fd9448dc4571cd8924975&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.2260 to 1.2300
Refractive index n20/D 1.5220 to 1.5260

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR