Stampa…

Ethyl phenylcyanoacetate ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

P1255-25G P1255-5G
TCIAP1255-25GEA 144 EUR
TCIAP1255-25G TCIAP1255-5G
Ethyl phenylcyanoacetate ≥95.0% (by GC)
Ethyl phenylcyanoacetate
Formula: C₁₁H₁₁NO₂
Peso molecolare: 189,21 g/mol
Punto di ebollizione: 275°C
MDL Number: MFCD00001866
CAS Number: 4553-07-5
EINECS: 224-921-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a32814c1dc81e065909ec0cb1abdaa3fc569face&vid=a32814c1dc81e065909ec0cb1abdaa3fc569face&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR