Tetrafluoroisophthalic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Perfluoroisophthalic acid, 2,4,5,6-Tetrafluoroisophthalic acid
Formula:
C₈H₂F₄O₄ Peso molecolare: 238.0927 g/mol |
MDL Number:
MFCD00042379 CAS Number: 1551-39-9 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 93.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 208.0 to 214.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...