3-Methoxy-4-methylbenzonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
M1977-1G
M1977-5G
TCIAM1977-1GEA
17
EUR
TCIAM1977-1G
TCIAM1977-5G
3-Methoxy-4-methylbenzonitrile ≥98.0% (by GC)
3-Methoxy-4-methylbenzonitrile
Formula:
C₉H₉NO Peso molecolare: 147,1739 g/mol |
MDL Number:
MFCD03428590 CAS Number: 3556-60-3 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...